Assay Details
| Gene Symbol: PHT1;6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123700.1 | NP_199148.1 | 1 | 1493 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHT1;6| Gene Symbol: PHT1;6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123700.1 | NP_199148.1 | 1 | 1493 | 75 | |
Set Membership: |
|