Assay Details
| Gene Symbol: AT5G43670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123733.2 | NP_568626.1 | 1 - 2 | 980 | 96 | |
| GenBank mRNA | BT003332.1 | - | 1 - 2 | 924 | 96 | |
| BT010350.1 | - | 1 - 2 | 723 | 96 | ||
More Information
Set Membership: |
|

AT5G43670
Genomic Map