Assay Details
| Gene Symbol: AT5G43745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123741.3 | NP_568628.1 | 17 - 18 | 2067 | 67 | |
| GenBank mRNA | AY052350.1 | - | 17 - 18 | 1982 | 67 | |
| AY059653.1 | - | 17 - 18 | 1970 | 67 | ||
More Information
Set Membership: |
|

AT5G43745
Genomic Map