Assay Details
| Gene Symbol: TOM22-V | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123764.3 | NP_199210.1 | 1 | 481 | 112 | |
| GenBank mRNA | AY052270.1 | - | 1 | 459 | 112 | |
| AY088551.1 | - | 1 | 462 | 112 | ||
More Information
Set Membership: |
|

TOM22-V
Genomic Map