Assay Details
| Gene Symbol: CESA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344528.1 | NP_001332727.1 | 10 - 11 | 2977 | 68 | |
| NM_001344529.1 | NP_001332726.1 | 6 - 7 | 2044 | 68 | ||
| NM_123770.4 | NP_199216.2 | 11 - 12 | 2890 | 68 | ||
| GenBank mRNA | AK228561.1 | - | 11 - 12 | 2638 | 68 | |
| BT005710.1 | - | 11 - 12 | 2638 | 68 | ||
| BT006111.1 | - | 11 - 12 | 2567 | 68 | ||
| KJ676989.1 | - | 11 - 12 | 2567 | 68 | ||
More Information
Set Membership: |
|

CESA4
Genomic Map