Assay Details
| Gene Symbol: CESA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123770.4 | NP_199216.2 | 3 - 4 | 666 | 138 | |
| GenBank mRNA | AK228561.1 | - | 3 - 4 | 414 | 138 | |
| BT005710.1 | - | 3 - 4 | 414 | 138 | ||
| BT006111.1 | - | 3 - 4 | 343 | 138 | ||
| KJ676989.1 | - | 3 - 4 | 343 | 138 | ||
More Information
Set Membership: |
|

CESA4
Genomic Map