Assay Details
| Gene Symbol: CESA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344528.1 | NP_001332727.1 | 3 - 4 | 831 | 65 | |
| NM_123770.4 | NP_199216.2 | 4 - 5 | 744 | 65 | ||
| GenBank mRNA | AK228561.1 | - | 4 - 5 | 492 | 65 | |
| BT005710.1 | - | 4 - 5 | 492 | 65 | ||
| BT006111.1 | - | 4 - 5 | 421 | 65 | ||
| KJ676989.1 | - | 4 - 5 | 421 | 65 | ||
More Information
Set Membership: |
|

CESA4
Genomic Map