Assay Details
| Gene Symbol: CAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123774.4 | NP_199220.1 | 1 - 2 | 245 | 121 | |
| GenBank mRNA | AF093753.1 | - | 1 - 2 | 75 | 121 | |
| AF135155.1 | - | 1 - 2 | 123 | 121 | ||
| AY039951.1 | - | 1 - 2 | 193 | 121 | ||
| AY048257.1 | - | 1 - 2 | 184 | 121 | ||
| AY079384.1 | - | 1 - 2 | 75 | 121 | ||
More Information
Set Membership: |
|

CAD1
Genomic Map