Assay Details
| Gene Symbol: CAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344531.1 | NP_001332549.1 | 2 - 3 | 672 | 80 | |
| NM_123774.4 | NP_199220.1 | 2 - 3 | 402 | 80 | ||
| GenBank mRNA | AF093753.1 | - | 2 - 3 | 232 | 80 | |
| AF135155.1 | - | 2 - 3 | 280 | 80 | ||
| AY039951.1 | - | 2 - 3 | 350 | 80 | ||
| AY048257.1 | - | 2 - 3 | 341 | 80 | ||
| AY079384.1 | - | 2 - 3 | 232 | 80 | ||
More Information
Set Membership: |
|

CAD1
Genomic Map