Assay Details
| Gene Symbol: ABCI21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036931.1 | NP_001032008.1 | 1 - 2 | 222 | 79 | |
| NM_001085235.1 | NP_001078704.1 | 1 - 2 | 222 | 79 | ||
| NM_123778.3 | NP_199224.1 | 1 - 2 | 248 | 79 | ||
| GenBank mRNA | AF127664.1 | - | 1 - 2 | 152 | 79 | |
| AK228176.1 | - | 1 - 2 | 209 | 79 | ||
| BT006201.1 | - | 1 - 2 | 152 | 79 | ||
More Information
Set Membership: |
|

ABCI21
Genomic Map