Assay Details
| Gene Symbol: ABCI21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036931.1 | NP_001032008.1 | 2 - 3 | 389 | 104 | |
| NM_001085235.1 | NP_001078704.1 | 2 - 3 | 389 | 104 | ||
| NM_001344534.1 | NP_001332312.1 | 2 - 3 | 567 | 104 | ||
| NM_123778.3 | NP_199224.1 | 2 - 3 | 415 | 104 | ||
| GenBank mRNA | AF127664.1 | - | 2 - 3 | 319 | 104 | |
| AK228176.1 | - | 2 - 3 | 376 | 104 | ||
| BT006201.1 | - | 2 - 3 | 319 | 104 | ||
More Information
Set Membership: |
|

ABCI21
Genomic Map