Assay Details
| Gene Symbol: ABCI21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036931.1 | NP_001032008.1 | 4 - 5 | 565 | 79 | |
| NM_001344534.1 | NP_001332312.1 | 4 - 5 | 743 | 79 | ||
| NM_123778.3 | NP_199224.1 | 4 - 5 | 591 | 79 | ||
| GenBank mRNA | AF127664.1 | - | 4 - 5 | 495 | 79 | |
| AK228176.1 | - | 4 - 5 | 552 | 79 | ||
| BT006201.1 | - | 4 - 5 | 495 | 79 | ||
More Information
Set Membership: |
|

ABCI21
Genomic Map