Assay Details
| Gene Symbol: AT5G44550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123822.4 | NP_199268.1 | 1 - 2 | 320 | 71 | |
| GenBank mRNA | AY065079.1 | - | 1 - 2 | 245 | 71 | |
| AY114543.1 | - | 1 - 2 | 211 | 71 | ||
More Information
Set Membership: |
|

AT5G44550
Genomic Map