Assay Details
| Gene Symbol: AT5G44680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123835.4 | NP_199281.1 | 1 - 2 | 704 | 87 | |
| GenBank mRNA | BT003900.1 | - | 1 - 2 | 572 | 87 | |
| BT004932.1 | - | 1 - 2 | 520 | 87 | ||
More Information
Set Membership: |
|

AT5G44680
Genomic Map