Assay Details
| Gene Symbol: RAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123847.3 | NP_199292.1 | 1 - 2 | 654 | 77 | |
| GenBank mRNA | AF082565.1 | - | 1 - 2 | 465 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAN1| Gene Symbol: RAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123847.3 | NP_199292.1 | 1 - 2 | 654 | 77 | |
| GenBank mRNA | AF082565.1 | - | 1 - 2 | 465 | 77 | |
Set Membership: |
|