Assay Details
| Gene Symbol: RAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123847.3 | NP_199292.1 | 4 - 5 | 2016 | 80 | |
| GenBank mRNA | AF082565.1 | - | 4 - 5 | 1827 | 80 | |
More Information
Set Membership: |
|

RAN1
Genomic Map