Assay Details
| Gene Symbol: RAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123847.3 | NP_199292.1 | 6 - 7 | 2418 | 65 | |
| GenBank mRNA | AF082565.1 | - | 6 - 7 | 2229 | 65 | |
More Information
Set Membership: |
|

RAN1
Genomic Map