Assay Details
| Gene Symbol: RAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123847.3 | NP_199292.1 | 7 - 8 | 2616 | 85 | |
| GenBank mRNA | AF082565.1 | - | 7 - 8 | 2427 | 85 | |
More Information
Set Membership: |
|

RAN1
Genomic Map