Assay Details
| Gene Symbol: BMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344606.1 | NP_001330613.1 | 1 - 2 | 590 | 96 | |
| NM_123898.3 | NP_199343.1 | 1 - 2 | 505 | 96 | ||
| GenBank mRNA | AK117140.1 | - | 1 - 2 | 502 | 96 | |
| BT006482.1 | - | 1 - 2 | 479 | 96 | ||
More Information
Set Membership: |
|

BMY2
Genomic Map