Assay Details
| Gene Symbol: BMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036937.1 | NP_001032014.1 | 4 - 5 | 1008 | 93 | |
| NM_001344606.1 | NP_001330613.1 | 4 - 5 | 1097 | 93 | ||
| NM_123898.3 | NP_199343.1 | 4 - 5 | 1012 | 93 | ||
| GenBank mRNA | AK117140.1 | - | 4 - 5 | 1009 | 93 | |
| BT006482.1 | - | 4 - 5 | 986 | 93 | ||
More Information
Set Membership: |
|

BMY2
Genomic Map