Assay Details
| Gene Symbol: BMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036937.1 | NP_001032014.1 | 9 - 10 | 2065 | 140 | |
| NM_001344606.1 | NP_001330613.1 | 9 - 10 | 2154 | 140 | ||
| NM_123898.3 | NP_199343.1 | 9 - 10 | 2069 | 140 | ||
| GenBank mRNA | AK117140.1 | - | 9 - 10 | 2066 | 140 | |
More Information
Set Membership: |
|

BMY2
Genomic Map