Assay Details
| Gene Symbol: CLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123907.4 | NP_568644.1 | 2 - 3 | 996 | 89 | |
| GenBank mRNA | AY042832.1 | - | 2 - 3 | 944 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLPP4| Gene Symbol: CLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123907.4 | NP_568644.1 | 2 - 3 | 996 | 89 | |
| GenBank mRNA | AY042832.1 | - | 2 - 3 | 944 | 89 | |
Set Membership: |
|