Assay Details
| Gene Symbol: GAS41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203546.1 | NP_001190475.1 | 5 - 6 | 831 | 72 | |
| NM_123928.4 | NP_199373.1 | 5 - 6 | 986 | 72 | ||
| GenBank mRNA | AY072123.1 | - | 5 - 6 | 882 | 72 | |
| AY096486.1 | - | 5 - 6 | 725 | 72 | ||
| AY463617.1 | - | 5 - 6 | 725 | 72 | ||
More Information
Set Membership: |
|

GAS41
Genomic Map