Assay Details
| Gene Symbol: CIPK20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123950.4 | NP_199394.1 | 1 | 1663 | 83 | |
| GenBank mRNA | AK221510.1 | - | 1 | 1414 | 83 | |
| AY035225.1 | - | 1 | 1271 | 83 | ||
| BT028970.1 | - | 1 | 1271 | 83 | ||
More Information
Set Membership: |
|

CIPK20
Genomic Map