Assay Details
| Gene Symbol: NUDT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125908.1 | NP_001119380.1 | 1 - 2 | 310 | 120 | |
| NM_001344651.1 | NP_001331230.1 | 1 - 2 | 295 | 120 | ||
| NM_123962.3 | NP_199406.1 | 1 - 2 | 295 | 120 | ||
More Information
Set Membership: |
|

NUDT11
Genomic Map