Assay Details
| Gene Symbol: PTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123973.4 | NP_199417.1 | 2 - 3 | 451 | 91 | |
| GenBank mRNA | AY150511.1 | - | 2 - 3 | 331 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTR3| Gene Symbol: PTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123973.4 | NP_199417.1 | 2 - 3 | 451 | 91 | |
| GenBank mRNA | AY150511.1 | - | 2 - 3 | 331 | 91 | |
Set Membership: |
|