Assay Details
| Gene Symbol: PTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344656.1 | NP_001331060.1 | 1 - 2 | 782 | 65 | |
| NM_123973.4 | NP_199417.1 | 3 - 4 | 1012 | 65 | ||
| GenBank mRNA | AY150511.1 | - | 3 - 4 | 892 | 65 | |
More Information
Set Membership: |
|

PTR3
Genomic Map