Assay Details
| Gene Symbol: DELTA-OAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123987.4 | NP_199430.1 | 1 - 2 | 630 | 86 | |
| GenBank mRNA | BT023421.1 | - | 1 - 2 | 170 | 86 | |
| BT029160.1 | - | 1 - 2 | 157 | 86 | ||
More Information
Set Membership: |
|

DELTA-OAT
Genomic Map