Assay Details
| Gene Symbol: MCM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344668.1 | NP_001332359.1 | 9 - 10 | 1604 | 62 | |
| NM_001344669.1 | NP_001332360.1 | 9 - 10 | 1604 | 62 | ||
| NM_001344670.1 | NP_001332361.1 | 9 - 10 | 1607 | 62 | ||
| NM_123997.3 | NP_199440.1 | 9 - 10 | 1604 | 62 | ||
More Information
Set Membership: |
|

MCM3
Genomic Map