Assay Details
| Gene Symbol: KCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124007.3 | NP_199449.1 | 1 - 2 | 1306 | 110 | |
| GenBank mRNA | DQ447044.1 | - | 1 - 2 | 1156 | 110 | |
More Information
Set Membership: |
|

KCO2
Genomic Map