Assay Details
| Gene Symbol: SSP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344678.1 | NP_001332006.1 | 2 - 3 | 394 | 71 | |
| NM_124011.4 | NP_199453.2 | 2 - 3 | 397 | 71 | ||
| GenBank mRNA | AY074261.1 | - | 2 - 3 | 394 | 71 | |
| AY096737.1 | - | 1 - 2 | 166 | 71 | ||
More Information
Set Membership: |
|

SSP4
Genomic Map