Assay Details
| Gene Symbol: SSP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125911.1 | NP_001119383.1 | 3 - 4 | 524 | 109 | |
| NM_001344678.1 | NP_001332006.1 | 3 - 4 | 515 | 109 | ||
| NM_124011.4 | NP_199453.2 | 3 - 4 | 518 | 109 | ||
| GenBank mRNA | AY074261.1 | - | 3 - 4 | 515 | 109 | |
| AY096737.1 | - | 2 - 3 | 287 | 109 | ||
More Information
Set Membership: |
|

SSP4
Genomic Map