Assay Details
| Gene Symbol: SSP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125911.1 | NP_001119383.1 | 5 - 6 | 1100 | 88 | |
| NM_001344678.1 | NP_001332006.1 | 5 - 6 | 1091 | 88 | ||
| NM_124011.4 | NP_199453.2 | 5 - 6 | 1094 | 88 | ||
| GenBank mRNA | AY074261.1 | - | 5 - 6 | 1091 | 88 | |
| AY096737.1 | - | 4 - 5 | 863 | 88 | ||
More Information
Set Membership: |
|

SSP4
Genomic Map