Assay Details
| Gene Symbol: AT5G46420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124012.3 | NP_568663.2 | 5 - 6 | 1398 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G46420| Gene Symbol: AT5G46420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124012.3 | NP_568663.2 | 5 - 6 | 1398 | 77 | |
Set Membership: |
|