Assay Details
| Gene Symbol: BSK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124027.3 | NP_199469.1 | 2 - 3 | 603 | 116 | |
| GenBank mRNA | BT015774.1 | - | 2 - 3 | 294 | 116 | |
More Information
Set Membership: |
|

BSK2
Genomic Map