Assay Details
| Gene Symbol: BSK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124027.3 | NP_199469.1 | 4 - 5 | 983 | 146 | |
| GenBank mRNA | BT015774.1 | - | 4 - 5 | 674 | 146 | |
More Information
Set Membership: |
|

BSK2
Genomic Map