Assay Details
| Gene Symbol: TRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124040.5 | NP_199482.1 | 1 - 2 | 689 | 125 | |
| GenBank mRNA | AK226278.1 | - | 1 - 2 | 616 | 125 | |
| BT020321.1 | - | 1 - 2 | 527 | 125 | ||
More Information
Set Membership: |
|

TRN2
Genomic Map