Assay Details
| Gene Symbol: HB-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344703.1 | NP_001318750.1 | 10 - 11 | 2125 | 101 | |
| NM_001344704.1 | NP_001330010.1 | 10 - 11 | 2203 | 101 | ||
| NM_124059.4 | NP_199499.3 | 9 - 10 | 2261 | 101 | ||
More Information
Set Membership: |
|

HB-7
Genomic Map