Assay Details
| Gene Symbol: AT5G46890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124060.3 | NP_199500.1 | 1 | 572 | 152 | |
| GenBank mRNA | AK117543.1 | - | 1 | 550 | 152 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G46890| Gene Symbol: AT5G46890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124060.3 | NP_199500.1 | 1 | 572 | 152 | |
| GenBank mRNA | AK117543.1 | - | 1 | 550 | 152 | |
Set Membership: |
|