Assay Details
| Gene Symbol: BI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124083.3 | NP_199523.1 | 2 - 3 | 364 | 104 | |
| GenBank mRNA | AB025927.2 | - | 2 - 3 | 295 | 104 | |
| AF208124.1 | - | 2 - 3 | 290 | 104 | ||
| AY091134.1 | - | 2 - 3 | 281 | 104 | ||
| AY114059.1 | - | 2 - 3 | 245 | 104 | ||
More Information
Set Membership: |
|

BI1
Genomic Map