Assay Details
| Gene Symbol: BI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124083.3 | NP_199523.1 | 3 - 4 | 469 | 66 | |
| GenBank mRNA | AB025927.2 | - | 3 - 4 | 400 | 66 | |
| AF208124.1 | - | 3 - 4 | 395 | 66 | ||
| AY091134.1 | - | 3 - 4 | 386 | 66 | ||
| AY114059.1 | - | 3 - 4 | 350 | 66 | ||
More Information
Set Membership: |
|

BI1
Genomic Map