Assay Details
| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124103.4 | NP_568681.1 | 3 - 4 | 373 | 70 | |
| GenBank mRNA | AY045963.1 | - | 3 - 4 | 273 | 70 | |
| AY091354.1 | - | 2 - 3 | 176 | 70 | ||
More Information
Set Membership: |
|

RPS19
Genomic Map