Assay Details
| Gene Symbol: mtACP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085257.1 | NP_001078726.1 | 1 - 2 | 453 | 103 | |
| NM_001344735.1 | NP_001331779.1 | 2 - 3 | 381 | 103 | ||
| NM_124136.5 | NP_199574.1 | 2 - 3 | 243 | 103 | ||
| GenBank mRNA | AY063951.1 | - | 2 - 3 | 235 | 103 | |
| AY096717.1 | - | 1 - 2 | 200 | 103 | ||
More Information
Set Membership: |
|

mtACP3
Genomic Map