Assay Details
| Gene Symbol: NUDT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344736.1 | NP_001331651.1 | 1 - 2 | 680 | 76 | |
| NM_001344737.1 | NP_001331652.1 | 2 - 3 | 361 | 76 | ||
| NM_001344738.1 | NP_001331649.1 | 1 - 2 | 307 | 76 | ||
| NM_001344739.1 | NP_001331650.1 | 1 - 2 | 240 | 76 | ||
| NM_124139.4 | NP_568687.1 | 2 - 3 | 411 | 76 | ||
| GenBank mRNA | AY042806.1 | - | 2 - 3 | 245 | 76 | |
| AY064646.1 | - | 1 - 2 | 169 | 76 | ||
More Information
Set Membership: |
|

NUDT2
Genomic Map