Assay Details
| Gene Symbol: ERF1-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124162.4 | NP_199599.1 | 2 - 3 | 1543 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERF1-1| Gene Symbol: ERF1-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124162.4 | NP_199599.1 | 2 - 3 | 1543 | 82 | |
Set Membership: |
|