Assay Details
| Gene Symbol: AT5G48020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124176.3 | NP_568689.1 | 3 - 4 | 689 | 82 | |
| GenBank mRNA | AK227707.1 | - | 3 - 4 | 630 | 82 | |
| AY087978.1 | - | 3 - 4 | 675 | 82 | ||
| BT008313.1 | - | 3 - 4 | 508 | 82 | ||
More Information
Set Membership: |
|

AT5G48020
Genomic Map