Assay Details
| Gene Symbol: MET18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344759.1 | NP_001318759.1 | 10 - 11 | 1668 | 82 | |
| NM_001344760.1 | NP_001330951.1 | 10 - 11 | 1641 | 82 | ||
| NM_124186.5 | NP_199623.2 | 10 - 11 | 1660 | 82 | ||
| GenBank mRNA | AK226792.1 | - | 10 - 11 | 1628 | 82 | |
More Information
Set Membership: |
|

MET18
Genomic Map