Assay Details
| Gene Symbol: MET18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344759.1 | NP_001318759.1 | 1 - 2 | 196 | 153 | |
| NM_001344760.1 | NP_001330951.1 | 1 - 2 | 169 | 153 | ||
| NM_124186.5 | NP_199623.2 | 1 - 2 | 188 | 153 | ||
| GenBank mRNA | AK226792.1 | - | 1 - 2 | 156 | 153 | |
More Information
Set Membership: |
|

MET18
Genomic Map