Assay Details
| Gene Symbol: AtSPO22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124214.7 | NP_199650.3 | 1 - 2 | 1405 | 148 | |
| GenBank mRNA | AB354634.1 | - | 1 - 2 | 1363 | 148 | |
More Information
Set Membership: |
|

AtSPO22
Genomic Map