Assay Details
| Gene Symbol: AT5G48490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124225.2 | NP_199660.1 | 1 | 301 | 65 | |
| GenBank mRNA | AY087218.1 | - | 1 | 220 | 65 | |
| BT025701.1 | - | 1 | 176 | 65 | ||
More Information
Set Membership: |
|

AT5G48490
Genomic Map